Standard Cell Factories now configurable in the Single Use Assembly Configurator Tool!
Nunc™ EasyFill™ Cell Factory™ System
Actual product may vary
Nunc™ EasyFill™ Cell Factory™ System
Nunc™ EasyFill™ Cell Factory™ System
Nunc™ EasyFill™ Cell Factory™ System
Nunc™ EasyFill™ Cell Factory™ System
Nunc™ EasyFill™ Cell Factory™ System
Nunc™ EasyFill™ Cell Factory™ System
Nunc™ EasyFill™ Cell Factory™ System
Thermo Scientific™
Nunc™ EasyFill™ Cell Factory™ System
Green features
Save valuable space with Thermo Scientific™ Nunc™ EasyFill™ Cell Factory system. Each 10-layer, single-use system holds the equivalent of 36 T175 flasks.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberTray LayersCulture AreaNo. per Case
1400001632 cm²6/Cs.
14025021264 cm²6/Cs.
14036042528 cm²4/Cs.
140400106320 cm²2/Cs.
140410106320 cm²6/Cs.
1404404025,280 cm²2/Cs.
Catalog number 140000
Price (USD)
602.00
Case of 6
Tray Layers:
1
Culture Area:
632 cm²
No. per Case:
6/Cs.
Price (USD)
602.00
Case of 6
Save valuable space with Thermo Scientific™ Nunc™ EasyFill™ Cell Factory system. Each 10-layer, single-use system holds the equivalent of 36 T175 flasks. The linear multi-layer cell culture format is easy to use and promotes easy scalability, ideal for both research and commercial-scale applications.
Features:
  • 'Out of the box' system
  • Large, vented screw closure
  • Versatile port design for pouring and aseptic filling
Benefits:
  • Fill and empty five times faster than T175 flasks
  • No accessories required with EasyFill Cell Factory systems
To Get Started:
Simply pour the media directly into the EasyFill Cell Factory system's large opening.
Warranty: 90 days
WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Closure TypeVented
DescriptionEasyFill Cell Factory
Green FeaturesISCC PLUS
No. per Case6/Cs.
No. per Pack1 Ea.
Tray Layers1
Culture Area632 cm²
SterilitySterile
Unit SizeCase of 6