Search
Extractables and leachables (E&L) testing is critical for ensuring the safety and compliance of pharmaceutical packaging and medical devices. Key applications in E&L analysis include: non-volatile compound workflows, volatile compound workflows, PFAS detection in packaging and medical devices, and elemental impurity analysis. Leveraging Thermo Fisher Scientific’s innovative instrumentation—such as high-resolution mass spectrometry (HRAM), gas chromatography-mass spectrometry (GC-MS), and inductively coupled plasma-mass spectrometry (ICP-MS)—our solutions provide the sensitivity, accuracy, and regulatory compliance needed for robust E&L workflows.
Non-volatile compound workflows identify high-molecular-weight, thermally stable extractables from packaging, medical devices, and biopharmaceutical components. Using liquid extraction techniques like accelerated solvent extraction (ASE) and LC-MS or UHPLC-HRMS, these workflows detect polymeric additives, oligomers, and degradation products. Thermo Scientific Orbitrap HRMS and Thermo Scientific Vanquish UHPLC systems provide high sensitivity and compliance, helping to ensure product safety and regulatory adherence.
Per- and polyfluoroalkyl substances (PFAS) are widely used in pharmaceutical packaging and medical devices due to their durability and chemical resistance, but their potential to leach into drug products raises safety concerns. Detecting PFAS requires sensitive analytical techniques such as LC-MS/MS and high-resolution mass spectrometry (HRMS) to identify and quantify these persistent contaminants at trace levels. Thermo Scientific Orbitrap HRMS and triple quadrupole LC-MS systems offer high precision and regulatory-compliant workflows for PFAS analysis, helping to ensure product safety and compliance with evolving guidelines.
Volatile extractables, such as residual solvents and monomers, can migrate from packaging and medical devices, posing risks to drug stability. HS-GC-MS, using Thermo Scientific TriPlus 500 Headspace Autosampler and Thermo Scientific Orbitrap GC-MS, enables sensitive, high-resolution detection to help ensure compliance and product safety.
Elemental impurities from packaging, manufacturing components, or drug formulations can pose toxicity risks and require strict regulatory compliance. Inductively coupled plasma mass spectrometry (ICP-MS) and inductively coupled plasma optical emission spectroscopy (ICP-OES) are the gold standard for detecting trace metals in pharmaceutical products. Thermo Scientific iCAP ICP-MS OES systems provide high sensitivity and accuracy, helping to ensure compliance with ICH Q3D guidelines for elemental impurity analysis in E&L studies.
For Research Use Only. Not for use in diagnostic procedures.