Search
Peptide array analysis was used to confirm the specificity of this antibody for its intended modification. Histone H3 (AcK9) Oligoclonal Antibody (Cat. No. 710293) was applied at a dilution of 1:3,000 to a MODified? Histone Peptide Array (Catalog No. 13001). The arrays were scanned with ArrayAnalysis Software and the results plotted. Specificity data is shown for the most reactive peptides.