PLA2R Antibody (MA1-212) in IHC
Immunohistochemistry was performed on human kidney tissue. Tissue was deparaffinized with xylene, followed by rehydration in sequential washes of 100% ethanol, 95% ethanol, 80% ethanol, and water. To expose target proteins, antigen retrieval was performed using 10mM sodium citrate (pH 6.0) and heated for 20 min. Following antigen retrieval, tissues were blocked in a 10% goat serum (Product ## 31872) in wash buffer solution for 30-60 minutes at room temperature. Tissue was then probed with a PLA2R Mouse Monoclonal antibody (Product ## MA1-212) at a dilution of 1:200 in 10% goat serum in wash buffer for at least one hour in a humidified chamber. Negative control tissue received no primary antibody. Tissues were washed extensively with PBST, and detection was performed using a goat anti-mouse IgG-HRP secondary antibody at a dilution of 1:500 followed by colorimetric detection using metal enhanced DAB. Tissues were then counterstained with hematoxylin and prepped for mounting and imaging.
Immunohistochemistry was performed on human kidney tissue. Tissue was deparaffinized with xylene, followed by rehydration in sequential washes of 100% ethanol, 95% ethanol, 80% ethanol, and water. To expose target proteins, antigen retrieval was performed using 10mM sodium citrate (pH 6.0) and heated for 20 min. Following antigen retrieval, tissues were blocked in a 10% goat serum (Product ## 31872) in wash buffer solution for 30-60 minutes at room temperature. Tissue was then probed with a PLA2R Mouse Monoclonal antibody (Product ## MA1-212) at a dilution of 1:200 in 10% goat serum in wash buffer for at least one hour in a humidified chamber. Negative control tissue received no primary antibody. Tissues were washed extensively with PBST, and detection was performed using a goat anti-mouse IgG-HRP secondary antibody at a dilution of 1:500 followed by colorimetric detection using metal enhanced DAB. Tissues were then counterstained with hematoxylin and prepped for mounting and imaging.
Related Products
Related Images