Panel key benefits
  • Provides formulation diversity not found when testing catalog products individually.
  • Can increase the possibility that you will be able to narrow in on a nutrient composition that best supports your clone’s specific needs.
  • Protocol and study plan included to simplify your experimental design.
  • Technical support and consultation provided.
  • Analysis of results with recommended next steps included.
  • Access to additional testing, such as spent media analysis, to allow further optimization.
What’s new: Gibco GlycanMod Panel
Introducing the Gibco GlycanMod Panel—precision control for protein galactosylation
The GlycanMod Panel is designed to optimize monoclonal antibody (mAb) galactosylation, supporting consistent, high-quality protein production. These chemically defined formulations work across various cell lines and media, providing the flexibility needed to fine-tune N-glycan profiles and meet therapeutic goals.
GlycanMod Panel
With titratable, ready-to-use solutions, the GlycanMod Panel enables rapid modification of protein galactosylation without compromising cell growth or productivity. Whether for early-stage development or cGMP manufacturing, GlycanMod helps streamline your process, accelerating time-to-market while ensuring batch-to-batch consistency. Help achieve greater control over your glycosylation process, and see how the GlycanMod Panel can enrich the quality and efficacy of your biopharmaceuticals.
gibco-hek-media-bottles
Viral Vector HEK media and feed panel
The Gibco Viral Vector HEK Media and Feed Panel accelerates gene therapy development and production by providing media and feeds that cover broad variety of suspension HEK293 cells without being tied to specific transfection reagents or techniques. The ready-to-use formulations will help you identify an optimal formulation for your target cell line.
The importance of formulation diversity
With catalog products, limited formulation information is available, making it challenging to set up an evaluation that is truly diverse. The Gibco Media and Feed Panel Evaluations were developed with diversity in mind, providing you a panel of products that each have distinct formulation and nutrient levels, allowing you to quickly narrow in on the components driving increases in productivity and quality.
Unsure which service might be right for you? 
Try our Media by Design Services interactive selection guide. Select your modality and challenge, learn about options available for your specific needs.
Services Selector