Beta-Actin Loading Control Antibody (MA5-15739-D800) in WB
Western blot analysis of Beta-Actin was performed by loading 50 µg of various cell lysates per well onto a 4-20% Tris-HCl polyacrylamide gel. Proteins were transferred to a nitrocellulose membrane and blocked with SEA BLOCK (Product # 37527) for at least 1 hour. The membrane was probed with a DyLight 800-conjugated Beta-Actin monoclonal antibody (Product # MA5-15739-D800) at a dilution of 1:1000 for 1 hour at room temperature on a rocking platform and washed in TBS-0.1% Tween-20. Detection was performed using the LI-COR Odyssey.
Western blot analysis of Beta-Actin was performed by loading 50 µg of various cell lysates per well onto a 4-20% Tris-HCl polyacrylamide gel.   Proteins were transferred to a nitrocellulose membrane and blocked with SEA BLOCK (Product # 37527) for at least 1 hour.  The membrane was probed with a DyLight 800-conjugated Beta-Actin monoclonal antibody (Product # MA5-15739-D800) at a dilution of 1:1000 for 1 hour at room temperature on a rocking platform and washed in TBS-0.1% Tween-20.  Detection was performed using the LI-COR Odyssey.
Related Products
Related Images