Search
Simply click the general GC symptom that you are observing, then navigate the symptom details to define potential causes of your problems and receive appropriate solutions to resolve the problem.
Double peaks | |
|---|---|
| Injection speed too low. | Inject more rapidly in a smooth motion. |
| Wrong autosampler injection speed or mode. | Use a higher speed. |
Irregular, chair-shaped peaks | |
|---|---|
| Solvent flooding of column. | Increase the initial oven temperature. Reduce the injection volume (OC). Install a retention gap (OC). |
Retention times decreasing | |
|---|---|
| Stationary phase deteriorated by oxygen and/or water. | Use a carrier gas free of oxygen and water. |
| Stationary phase loss due to column bleeding. | Reduce the column temperature. |
Retention times increasing | |
|---|---|
| Increasing carrier leakage. | Check the septum and column connections. |
| Carrier gas supply running out. | Replace the bottle. |