Search
Quickly find high-purity metals, alloys, and materials for your application. Use the search tool to locate exactly what you need.
Filters
*Full color tiles represent pure elements for which we offer a range of products.
*Faded color tiles represent pure elements for which products are currently not available.
Trusted purity and quality
|
Fast, reliable delivery
|
![]() |
Competitive pricing at any scale
|
Drug research Gold nanoparticles are being used in the development of a new generation of biologically active compounds when added to plastics, coatings, nanofibers, and textiles. Nanoparticles cross-linked with collagen are used in research on aging and damaged skin. |
|
Technology Gold in an elemental form is used in the mounting of microprocessors, in memory chips, as connectors used to attach cables, as well as in GPS systems, calculators, and fuel cell applications. Gold nanoparticles are being explored for use as sensory probes, electronic conductors, and organic photovoltaics. |
|
Chemistry Supported gold catalysts are used in a wide range of chemical reactions, including carbon monoxide oxidation, catalytic combustion of hydrocarbons, hydrochlorination of ethyne, hydrogen peroxide formation, oxidation of glucose to gluconic acid, oxidative decomposition of dioxins, ozone decomposition, NOx reduction, epoxidation of olefins, and hydrogenation of alkynes. Gold alloy catalysts are used in the commercial production of vinyl acetate monomer (VAM). |
|
Aerospace Pure gold in one or the other form is used in space vehicle circuitry, as a lubricant between vehicle parts, and as a radiation reflector for stabilizing the temperature inside the space vehicle. |
Chemistry Finely divided palladium, such as palladium on carbon, is used as a heterogeneous catalyst in hydrogenation, dehydrogenation, and petroleum cracking. Combined with a variety of ligands (e.g., bis(triphenylphosphine)palladium(II) dichloride), palladium acts as a homogeneous catalyst in the synthesis of fine chemicals. The 2010 Nobel Prize in chemistry recognized the significance of palladium reagents as catalysts in the synthetically important Heck, Negishi, and Suzuki cross-coupling reactions. |
|
Emission control devices Over 57% of the palladium supply is used in the catalytic converters found in more than 98% of all new vehicles. Palladium in these devices play a crucial role in converting harmful gases emitted by gasoline engines, such as carbon monoxide, hydrocarbon, and oxides of nitrogen into less harmful carbon dioxide, nitrogen, and water vapor. |
|
Jewelry Palladium jewelry is 95% pure and will not tarnish like sterling silver. It is also hypoallergenic, making it the metal of choice for sensitive skin. Its malleability allows designers to make very intricate pieces. Palladium is often used in white gold alloys, being lighter than platinum and purer than white gold, and it does not need to be plated with rhodium plating to keep it shiny. |
|
Electronics Because of its electrical conductivity and its durability, palladium is widely used in electronics. A small amount of the metal is used in the components of virtually every type of electronic device, from basic consumer products to complex military hardware. The metal is most commonly used in multilayer ceramic capacitors (MLCC) found in laptop computers and mobile phones. Smaller amounts of palladium are used in hybrid integrated circuits (HIC) and for plating connectors and lead frames. |
Chemistry The most common application of platinum is as a catalyst in chemical reactions. In industry, it is used in: the hydrogenation of benzene to produce cyclohexane, the raw material for nylon; fuel cells for the reduction of oxygen; the production of higher-octane gasoline; the manufacture of specialty silicones; and the hydrogenation of vegetable oils. Platinum and its alloys are used in the manufacture of crucibles and evaporating dishes for chemical analyses. In the research lab, organometallic complexes of platinum are used as reagents for the synthesis of many useful and interesting organic compounds. |
|
Emission control devices Records show that in 2014, 98 tons of platinum (about 45% of total platinum mined) were used in emission control devices called catalytic converters, which help convert over 90% of harmful elements like hydrocarbons, carbon monoxide, and oxides of nitrogen from gasoline engines into less harmful carbon dioxide, nitrogen, and water vapor. Over 98% of all new vehicles come with catalytic converters. |
|
Agriculture Platinum-rhodium catalysts are used in the manufacture of nitric acid, which in turn is used to produce the ammonia found in many liquid fertilizers for agricultural applications. |
|
Electronics Because of its unsurpassed metallurgical properties, platinum has been used in electrical contacts and in various electrode systems for many years. The microelectronics revolution that began in the 1970s opened up a range of new applications, including thin film technology (in integrated circuits as silicide ohmic and Schottky contacts and on-chip capacitors, as temperature sensors), thick film technology (as platinum/platinum alloy conductors and in resistance thermometers), and manufacturing (as crucibles for crystal growing, in thermocouples, and in glass-melting equipment for the manufacture of optical fibers). |
|
Aerospace Platinum is used in multiple ways in the manufacture of aircraft engine parts: in the coating of jet engine fuel nozzles and turbine blades used to protect against hot corrosion and oxidation, as a pinning wire to hold the ceramic cores within the aircraft turbine blade during casting, and in aerospace spark plugs and thermocouples. It is also used in the coating of missile cones. |
|
Jewlery The least reactive precious metal, known for its resistance to corrosion and oxidation at any temperature as well as for its ductility, malleability, and its silvery sheen, platinum is commonly used in jewelry. In 2010, 76 tons of platinum (about 31% of the platinum mined) was used in jewelry. |
Chemistry Rhodium is used in research and industrial laboratories primarily as a catalyst. It is preferable to the other platinum group catalysts in the reduction of nitrogen oxides to nitrogen and oxygen. Rhodium is also used to catalyze the reduction of benzene to cyclohexane as well as the addition of hydrosilanes to double bonds, an important step in the manufacture of certain silicone rubbers. In industry, it is used in the catalytic carbonylation of methanol to produce acetic acid by the Monsanto process. The coordination complex of rhodium(I) with tris(triphenylphosphine)chloride, known as Wilkinson's catalyst, is widely used in the hydrogenation of alkenes. The rhodium complex with BINAP(2,2′-bis(diphenylphosphino)-1,1′-binaphthyl) is used for chiral synthesis, as in the synthesis of methanol. Rhodium's ability to withstand high temperatures makes it a perfect material for laboratory crucibles. |
|
Emission control devices Approximately 87.2% of world rhodium production is used in catalytic converters, often together with palladium and/or platinum, to clean vehicle emissions. It has by far the highest activity for the removal of nitrogen oxides (NOx) from exhaust fumes, as well as very high activity for the oxidation of hydrocarbons and carbon monoxide and very good resistance to the poisons present in the exhaust stream. Its primary drawback, however, is its high cost. |
|
Jewlry and decoration Its brilliance and resistance to wear and tarnishing make rhodium a perfect material for jewelry. It is electroplated on white gold and platinum jewelry with a surface that guards against scratches and gives a reflective white appearance, preventing tarnishing during wear. It is also used in coating sterling silver to protect against tarnish from the silver sulfide produced from atmospheric hydrogen sulfide. Rhodium has also been used to signify elite status, when more commonly used metals such as silver, gold, or platinum were deemed insufficient, as when in 1979 the Guinness Book of World Records gave Paul McCartney a rhodium-plated disc for being history's all-time best-selling songwriter and recording artist. |
|
Industry Rhodium is used in the glass industry, mostly for production of fiberglass and flat-panel glass, and in the chemical industry, such as in producing some herbicides, as well as acids such as nitric acid and acetic acid. In automobile manufacturing, rhodium is used in the construction of headlight reflectors. Because of its rarity and its inertness against corrosion and most aggressive chemicals, rhodium is usually alloyed with platinum or palladium for use in high-temperature and corrosion-resistive coatings. Rhodium is sometimes used to cure silicones. Rhodium plated by either electroplating or evaporation is used in the manufacture of optical instruments as well as filters for mammography systems because of the characteristic X-rays it produces. Rhodium neutron detectors are used in nuclear reactors to measure neutron flux levels, providing an accurate 3D "picture" of reactivity and allowing fine tuning to determine the most economical consumption of nuclear fuel. |
|
Electrical Rhodium is often alloyed with platinum and iridium to make an oxidation-resistant metal that can withstand high temperatures. These alloys are used in furnace coils, thermocouple and resistance wires, and electrodes for aircraft spark plugs. It is also used in electrical contacts, where it is valued for its low electrical resistance, low and stable contact resistance, and great corrosion resistance. |
Chemical synthesis Many silver salts are used as precursors to other silver compounds and metallic silver, as well as reagents in organic reactions such as glycosylation. They also play a role in catalysis in chemical processes in industrial and professional settings. |
|
Energy Silver oxide batteries are used in small wearable devices and also have research applications in photovoltaics and fuel cells. |
|
Electronics Exhibiting superior bondability, solderability, and corrosion resistance, as well as durability and high conductivity, silver products are used in electrical contacts and printed electronics for mobile devices, displays, and touchscreens. |
|
Histology and proteomics Silver salt solutions are used extensively as stains in histology to visualize intra- and extracellular components, including proteins and DNA. |
|
Industrial water filtration Because of its antimicrobial properties, silver is used in the filtration and disinfection of industrial water and to prevent bacterial buildup in pools. Browse our silver nanoparticles available for your research in this field. |
Thermo Fisher Scientific offers a broad portfolio of high purity metals and materials, including pure elements, alloys, carbon and graphite products, ceramics, evaporation materials, optics, crystals, and specialty labware. With over 1,400 pure element products and hundreds of alloys and carbon materials available, we support advanced research and high-technology applications across industries.
Our pure metals and alloys are available in a wide range of purities, including ultra-high purity options up to 99.999% (metals basis). Purities are typically based on total metallic impurities (TMI), and select grades such as Thermo Scientific™ Puratronic™ and REacton™ include a detailed Certificate of Analysis (COA) outlining impurity levels and detection limits.
Yes. We provide a comprehensive selection of evaporation materials and sputtering targets for thin film deposition and crystal growth applications. Products are available in various purities and forms, including slugs, targets, and specialty configurations, designed to meet precise research and manufacturing specifications.
Our portfolio includes a wide range of carbon, graphite, and ceramic materials, such as carbon black, activated carbon, diamond powder, fullerene (C60), glassy carbon plates and rods, and high-temperature ceramic materials. These products are available in multiple forms and purities to support demanding laboratory and industrial applications.
Absolutely. For custom specifications, specialty fabrication, or large quantities, you can request a quote online. Request a quote now.
For Research Use Only. Not for use in diagnostic procedures.