Technical Resources - Media Formulations
18912 - PBS Tablets
Solid PBS tablet to be dissolved in 500ml distilled water.
  • Catalog Number(s)
ComponentsMolecular WeightConcentration (mg/L)mM
Inorganic Salts
Phosphate (as sodium phosphates)95.0950.010.0
Potassium Chloride (KCl)75.0201.02.68
Sodium Chloride (NaCl)58.08120.0140.0
参照
Related Products:
20012 - PBS, pH 7.2
Cat No: 20012019 , 20012027 , 20012043 , 20012050 , 20012068
70013 - PBS, 10x, pH 7.2
Cat No: 70013016 , 70013032 , 70013065 , 70013073
10010 - PBS, pH 7.4
Cat No: 10010001 , 10010002 , 10010015 , 10010023 , 10010031 , 10010049 , 10010056 , 10010072
70011 - PBS,10x, pH 7.4
Cat No: 70011036 , 70011044 , 70011051 , 70011069