Search
View additional product information for GeneChip™ Scanner 3000 7G Whole-Genome Association System - FAQs (00-0362)
1 product FAQs found
We have tested our assays in conditions between 13-37% humidity and obtained good results. Therefore, we recommend staying within this range.
Find additional tips, troubleshooting help, and resources within our Microarray Analysis Support Center.