Product Image
Invitrogen™
eBioscience™ Phytohemagglutinin-L (PHA-L) Solution (500X)
The Phytohemagglutinin-L (PHA-L) Solution (500X) is a ready to use solution of PHA-L in aqueous buffer. PHA is a mixtureRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
00-4977-93100 μL
00-4977-034 x 100 μL
Catalog number 00-4977-93
Price (USD)
95.65
Online Exclusive
103.00
Save 7.35 (7%)
Each
Quantity:
100 μL
Price (USD)
95.65
Online Exclusive
103.00
Save 7.35 (7%)
Each
The Phytohemagglutinin-L (PHA-L) Solution (500X) is a ready to use solution of PHA-L in aqueous buffer. PHA is a mixture of lectins isolated from the red kidney bean, Phaseolus vulgaris. PHA-L is the fraction of PHA that exhibits strong leukoagglutinating and mitogenic activity, but very little erythroagglutinating activity. This reagent is intended for use in in vitro activation of human and mouse leukocytes.
Reported Application
Functional Assays
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Product TypePhytohemagglutinin-L (PHA-L) Solution
Quantity100 μL
FormLiquid
Product LineeBioscience
Unit SizeEach