Product Image
Invitrogen™
eBioscience™ Protein Transport Inhibitor Cocktail (500X)
The Protein Transport Inhibitor Cocktail (500X) is a cocktail of Brefeldin A and Monensin. It is suitable for use inRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
00-4980-034 x 100 μL
00-4980-93100 μL
Catalog number 00-4980-03
Price (USD)
148.65
Online exclusive
160.00
Save 11.35
Each
Quantity:
4 x 100 μL
Price (USD)
148.65
Online exclusive
160.00
Save 11.35
Each
The Protein Transport Inhibitor Cocktail (500X) is a cocktail of Brefeldin A and Monensin. It is suitable for use in assays in which prevention of protein transport to the extracellular space is desired and has been specifically formulated for use as a negative control for the Cell Stimulation Cocktail (plus protein transport inhibitors) (500X) (cat. 00-4975). Addition of this cocktail to cells in culture inhibits the intracellular protein secretory/transport pathway resulting in the accumulation of secreted proteins in the lumen of the endoplasmic reticulum and in the Golgi apparatus. This accumulation can then be detected by intracellular staining and flow cytometric analysis or western blotting.
Reported Application
Flow Cytometric Analysis, Functional Assays, Cell Culture
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Quantity4 x 100 μL
Product LineeBioscience
TypeProtein Transport Inhibitor Cocktail
Unit SizeEach