Cadmium wire, 0.64mm (0.025in) dia, Puratronic™, 99.999% (metals basis)
Cadmium wire, 0.64mm (0.025in) dia, Puratronic™, 99.999% (metals basis)
Cadmium wire, 0.64mm (0.025in) dia, Puratronic™, 99.999% (metals basis)
Thermo Scientific Chemicals

Cadmium wire, 0.64mm (0.025in) dia, Puratronic™, 99.999% (metals basis)

CAS: 7440-43-9 | Cd | 112.41 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
3 m
15 m
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 000060.G4
also known as 000060-G4
Price (USD)
-
Quantity:
3 m
Request bulk or custom format
Chemical Identifiers
CAS7440-43-9
IUPAC Namecadmium
Molecular FormulaCd
InChI KeyBDOSMKKIYDKNTQ-UHFFFAOYSA-N
SMILES[Cd]
View more
SpecificationsSpecification SheetSpecification Sheet
FormWire, 0.64 mm (0.025 in.) dia
Assay from Suppliers CofAReport metals analysis
Total Metal Impurities≤0.001%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov
RUO – Research Use Only