Erbium pieces, 99.9% (REO)
Erbium pieces, 99.9% (REO)
Erbium pieces, 99.9% (REO)
Thermo Scientific Chemicals

Erbium pieces, 99.9% (REO)

CAS: 7440-52-0 | Er | 167.26 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Packaging:
View all
Glass Bottle under Argon
Catalog number 000111.09
also known as 000111-09
Price (USD)
123.65
Special offer
Online exclusive
Ends: 28-Dec-2026
145.00
Save 21.35
Each
Quantity:
10 g
Packaging:
Glass Bottle under Argon
Request bulk or custom format
Price (USD)
123.65
Special offer
Online exclusive
Ends: 28-Dec-2026
145.00
Save 21.35
Each
Chemical Identifiers
CAS7440-52-0
IUPAC Nameerbium
Molecular FormulaEr
InChI KeyUYAHIZSMUZPPFV-UHFFFAOYSA-N
SMILES[Er]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Rare Earth Oxide Impurities0.1% max.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only