Indium powder, -325 mesh, Puratronic™, 99.995% (metals basis)
Indium powder, -325 mesh, Puratronic™, 99.995% (metals basis)
Indium powder, -325 mesh, Puratronic™, 99.995% (metals basis)
Thermo Scientific Chemicals

Indium powder, -325 mesh, Puratronic™, 99.995% (metals basis)

CAS: 7440-74-6 | In | 114.82 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number 000151.06
also known as 000151-06
Price (USD)
157.65
Special offer
Online exclusive
Ends: 28-Dec-2026
185.00
Save 27.35
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
157.65
Special offer
Online exclusive
Ends: 28-Dec-2026
185.00
Save 27.35
Each
Chemical Identifiers
CAS7440-74-6
IUPAC Nameindium
Molecular FormulaIn
InChI KeyAPFVFJFRJDLVQX-UHFFFAOYSA-N
SMILES[In]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Silver-grey
Total Metal Impurities≤0.005%
Particle Size-325 mesh
FormPowder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only