Terbium ingot, 99.9% (REO), Ta <0.4%
Terbium ingot, 99.9% (REO), Ta <0.4%
Terbium ingot, 99.9% (REO), Ta <0.4%
Thermo Scientific Chemicals

Terbium ingot, 99.9% (REO), Ta <0.4%

CAS: 7440-27-9 | Tb | 158.93 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2 g
10 g
Catalog number 000320.04
also known as 000320-04
Price (USD)
96.65
Special offer
Online exclusive
Ends: 30-Jun-2026
114.00
Save 17.35 (15%)
Each
Quantity:
2 g
Request bulk or custom format
Price (USD)
96.65
Special offer
Online exclusive
Ends: 30-Jun-2026
114.00
Save 17.35 (15%)
Each
Chemical Identifiers
CAS7440-27-9
IUPAC Nameterbium
Molecular FormulaTb
InChI KeyGZCRRIHWUXGPOV-UHFFFAOYSA-N
SMILES[Tb]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Silver
Total Rare Earth Oxide Impurities0.1% max.
Impurity contentTa < 0.4%
FormIngot
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only