Terbium powder, -40 mesh, 99.9% (REO)
Terbium powder, -40 mesh, 99.9% (REO)
Terbium powder, -40 mesh, 99.9% (REO)
Thermo Scientific Chemicals

Terbium powder, -40 mesh, 99.9% (REO)

CAS: 7440-27-9 | Tb | 158.93 g/mol
Have Questions?
Catalog NumberQuantity
000322.03
also known as 000322-03
1 g
Catalog number 000322.03
also known as 000322-03
Price (USD)
-
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS7440-27-9
IUPAC Nameterbium
Molecular FormulaTb
InChI KeyGZCRRIHWUXGPOV-UHFFFAOYSA-N
SMILES[Tb]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Rare Earth Oxide Impurities0.1% max.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
  • Packaged in air stable poly bottle
RUO – Research Use Only