Titanium foil, 0.025mm (0.001in) thick, 99.94% (metals basis)
Titanium foil, 0.025mm (0.001in) thick, 99.94% (metals basis)
Titanium foil, 0.025mm (0.001in) thick, 99.94% (metals basis)
Thermo Scientific Chemicals

Titanium foil, 0.025mm (0.001in) thick, 99.94% (metals basis)

Form: Approx. 0.2g/25 x 75mm | CAS: 7440-32-6 | Ti | 47.87 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 x 75 mm
75 x 75 mm
150 x 150 mm
Catalog number 000360.FH
also known as 000360-FH
Price (USD)
148.65
Online Exclusive
165.00
Save 16.35 (10%)
Each
Quantity:
25 x 75 mm
Request bulk or custom format
Price (USD)
148.65
Online Exclusive
165.00
Save 16.35 (10%)
Each
Chemical Identifiers
CAS7440-32-6
IUPAC Nametitanium
Molecular FormulaTi
InChI KeyRTAQQCXQSZGOHL-UHFFFAOYSA-N
SMILES[Ti]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Silvery
Assay from Supplier's CofA≥99.94% (metals basis)
FormFoil, 0.025mm (0.001in) thick.
FormFoil, 0.025mm (0.001in) thick.
Assay from Supplier's CofA99.94% (metals basis)
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only