Zinc rod, 1.27cm (0.5in) dia, 99.9+% (metals basis)
Zinc rod, 1.27cm (0.5in) dia, 99.9+% (metals basis)
Zinc rod, 1.27cm (0.5in) dia, 99.9+% (metals basis)
Zinc rod, 1.27cm (0.5in) dia, 99.9+% (metals basis)
Zinc rod, 1.27cm (0.5in) dia, 99.9+% (metals basis)
Thermo Scientific Chemicals

Zinc rod, 1.27cm (0.5in) dia, 99.9+% (metals basis)

Form: approx. 9.03g/cm | CAS: 7440-66-6 | Zn | 65.38 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
15 cm
45 cm
90 cm
Catalog number 000422.BT
also known as 000422-BT
Price (USD)
59.40
Each
Add to cart
Quantity:
15 cm
Request bulk or custom format
Price (USD)
59.40
Each
Add to cart
Chemical Identifiers
CAS7440-66-6
IUPAC Namezinc
Molecular FormulaZn
InChI KeyHCHKCACWOHOZIP-UHFFFAOYSA-N
SMILES[Zn]
View more
SpecificationsSpecification SheetSpecification Sheet
FormRod, 1.27 cm dia.
Total Metal Impurities<0.1%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only