Tungsten wire, 0.075mm (0.003in) dia, 99.95% (metals basis)
Tungsten wire, 0.075mm (0.003in) dia, 99.95% (metals basis)
Tungsten wire, 0.075mm (0.003in) dia, 99.95% (metals basis)
Thermo Scientific Chemicals

Tungsten wire, 0.075mm (0.003in) dia, 99.95% (metals basis)

Form: approx. 15.2g/m | CAS: 7440-33-7 | W | 183.84 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 m
500 m
Catalog number 000457.H9
also known as 000457-H9
Price (USD)
289.65
Special offer
Online exclusive
Ends: 30-Jun-2026
340.00
Save 50.35 (15%)
Each
Quantity:
500 m
Request bulk or custom format
Price (USD)
289.65
Special offer
Online exclusive
Ends: 30-Jun-2026
340.00
Save 50.35 (15%)
Each
Chemical Identifiers
CAS7440-33-7
IUPAC Nametungsten
Molecular FormulaW
InChI KeyWFKWXMTUELFFGS-UHFFFAOYSA-N
SMILES[W]
View more
SpecificationsSpecification SheetSpecification Sheet
FormWire, 0.075mm dia.
Total Metal Impurities≤0.05%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only