Silver foil, 0.075mm (0.003in) thick, hard, 99.95% (metals basis)
Silver foil, 0.075mm (0.003in) thick, hard, 99.95% (metals basis)
Silver foil, 0.075mm (0.003in) thick, hard, 99.95% (metals basis)
Thermo Scientific Chemicals

Silver foil, 0.075mm (0.003in) thick, hard, 99.95% (metals basis)

CAS: 7440-22-4 | Ag | 107.87 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 x 150 mm
150 x 150 mm
Catalog number 000501.FS
also known as 000501-FS
Price (USD)
-
Quantity:
25 x 150 mm
Request bulk or custom format
Chemical Identifiers
CAS7440-22-4
IUPAC Namesilver
Molecular FormulaAg
InChI KeyBQCADISMDOOEFD-UHFFFAOYSA-N
SMILES[Ag]
View more
SpecificationsSpecification SheetSpecification Sheet
FormFoil, 0.075 mm thick
Total Metal Impurities≤0.05%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only