Praseodymium foil, 0.62mm (0.024 in.) thick, 99.5% (REO), Thermo Scientific Chemicals
Praseodymium foil, 0.62mm (0.024 in.) thick, 99.5% (REO), Thermo Scientific Chemicals
Praseodymium foil, 0.62mm (0.024 in.) thick, 99.5% (REO), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Praseodymium foil, 0.62mm (0.024 in.) thick, 99.5% (REO), Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantity
000607.FF
also known as 000607-FF
25 x 25 mm
Catalog number 000607.FF
also known as 000607-FF
Price (USD)
320.65
Special offer
Online exclusive
Ends: 30-Jun-2026
377.00
Save 56.35 (15%)
Each
Quantity:
25 x 25 mm
Request bulk or custom format
Price (USD)
320.65
Special offer
Online exclusive
Ends: 30-Jun-2026
377.00
Save 56.35 (15%)
Each
Chemical Identifiers
CAS7440-10-0
IUPAC Namepraseodymium
Molecular FormulaPr
InChI KeyPUDIUYLPXJFUGB-UHFFFAOYSA-N
SMILES[Pr]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Rare Earth Oxide Impurities0.5% max.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
  • Packaged under argon
RUO – Research Use Only