Copper powder, -625 mesh, APS 2-3.50 micron, 99% (metals basis)
Copper powder, -625 mesh, APS 2-3.50 micron, 99% (metals basis)
Copper powder, -625 mesh, APS 2-3.50 micron, 99% (metals basis)
Copper powder, -625 mesh, APS 2-3.50 micron, 99% (metals basis)
Thermo Scientific Chemicals

Copper powder, -625 mesh, APS 2-3.50 micron, 99% (metals basis)

CAS: 7440-50-8 | Cu | 63.55 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
250 g
Catalog number 000622.30
also known as 000622-30
Price (USD)
277.65
Online Exclusive
308.00
Save 30.35 (10%)
Each
Quantity:
250 g
Request bulk or custom format
Price (USD)
277.65
Online Exclusive
308.00
Save 30.35 (10%)
Each
Chemical Identifiers
CAS7440-50-8
IUPAC Namecopper
Molecular FormulaCu
InChI KeyRYGMFSIKBFXOCR-UHFFFAOYSA-N
SMILES[Cu]
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder, -625 mesh, APS 2-3.5 micron
Total Metal Impurities≤1.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only