Lutetium pieces, 99.9% (REO)
Lutetium pieces, 99.9% (REO)
Lutetium pieces, 99.9% (REO)
Thermo Scientific Chemicals

Lutetium pieces, 99.9% (REO)

CAS: 7439-94-3 | Lu | 174.97 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number 000628.03
also known as 000628-03
Price (USD)
223.65
Online Exclusive
248.00
Save 24.35 (10%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
223.65
Online Exclusive
248.00
Save 24.35 (10%)
Each
Chemical Identifiers
CAS7439-94-3
IUPAC Namelutetium
Molecular FormulaLu
InChI KeyOHSVLFRHMCKCQY-UHFFFAOYSA-N
SMILES[Lu]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Rare Earth Oxide Impurities0.1% max.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
  • Packaged in air-stable glass bottle
RUO – Research Use Only