Platinum foil, 0.635mm (0.025in) thick, 99.9% (metals basis)
Platinum foil, 0.635mm (0.025in) thick, 99.9% (metals basis)
Platinum foil, 0.635mm (0.025in) thick, 99.9% (metals basis)
Thermo Scientific Chemicals

Platinum foil, 0.635mm (0.025in) thick, 99.9% (metals basis)

CAS: 7440-06-4 | Pt | 195.08 g/mol
Have Questions?
Catalog NumberQuantity
000860.FF
also known as 000860-FF
25 x 25 mm
Catalog number 000860.FF
also known as 000860-FF
Price (USD)
3,299.00
Each
Quantity:
25 x 25 mm
Request bulk or custom format
Price (USD)
3,299.00
Each
Chemical Identifiers
CAS7440-06-4
IUPAC Nameplatinum
Molecular FormulaPt
InChI KeyBASFCYQUMIYNBI-UHFFFAOYSA-N
SMILES[Pt]
View more
SpecificationsSpecification SheetSpecification Sheet
FormFoil, 0.635mm (0.025in) thick.
Appearance (Color)Silver
Assay from Supplier's CofA≥99.9% (metals basis)
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only