Tantalum slug, 6.35mm (0.25 in.) dia. x 12.7mm (0.50 in.) length, 99.98% (metals basis), Thermo Scientific Chemicals
Tantalum slug, 6.35mm (0.25 in.) dia. x 12.7mm (0.50 in.) length, 99.98% (metals basis), Thermo Scientific Chemicals
Tantalum slug, 6.35mm (0.25 in.) dia. x 12.7mm (0.50 in.) length, 99.98% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Tantalum slug, 6.35mm (0.25 in.) dia. x 12.7mm (0.50 in.) length, 99.98% (metals basis), Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantity
000933.KJ
also known as 000933-KJ
5 Pc.
Catalog number 000933.KJ
also known as 000933-KJ
Price (USD)
148.65
Special offer
Online exclusive
Ends: 30-Jun-2026
175.00
Save 26.35 (15%)
Each
Quantity:
5 Pc.
Request bulk or custom format
Price (USD)
148.65
Special offer
Online exclusive
Ends: 30-Jun-2026
175.00
Save 26.35 (15%)
Each
Chemical Identifiers
CAS7440-25-7
IUPAC Nametantalum
Molecular FormulaTa
InChI KeyGUVRBAGPIYLISA-UHFFFAOYSA-N
SMILES[Ta]
View more
SpecificationsSpecification SheetSpecification Sheet
Assay from Supplier's CofA≥99.98% (metals basis)
FormSlug, 6.35mm (0.25in) diameter.
Appearance (Color)Silvery
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only