Product Image
Invitrogen™

TMB Chromogen Solution (for ELISA)

TMB (3,3', 5,5;-tetramethylbenzidine) chromogen solution may be used as a substrate for horseradish peroxidase (HRP) in ELISAs.Using TMBUse this reagentRead more
Have Questions?
Catalog NumberQuantity
002023500 mL
Catalog number 002023
Price (USD)
238.65
Online exclusive
254.00
Save 15.35
Each
Quantity:
500 mL
Request bulk or custom format
Price (USD)
238.65
Online exclusive
254.00
Save 15.35
Each

TMB (3,3', 5,5;-tetramethylbenzidine) chromogen solution may be used as a substrate for horseradish peroxidase (HRP) in ELISAs.

Using TMB

Use this reagent as supplied, no dilution or further preparation is required. In the presence of peroxidase (HRP, POD), TMB will be converted to a blue color that can be read at OD650 nm.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Product TypeTMB Solution
Quantity500 mL
Shipping ConditionWet Ice
SubstrateTMB
Substrate PropertiesChemical Substrate
Substrate TypeHRP (Horseradish Peroxidase) Substrate
Detection MethodColorimetric
Unit SizeEach
Contents & Storage
Store at 2°C to 8°C.