You need to enable JavaScript to run this app.
Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
New Products
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
Home
▸
Shop All Products
▸
Immunoassay Reagents and Kits
▸
Life Science Research ELISA Kits
▸
pNPP Solution
Invitrogen™
pNPP Solution
pNPP may be used as a substrate for alkaline phosphatase (AP) in enzyme
immunoassays.
Read more
Have Questions?
Catalog Number
Quantity
002212
100 ML
Catalog number
002212
Price (USD)
185.65
Online exclusive
193.00
Save 7.35
Each
Add to cart
Quantity:
100 ML
Price (USD)
185.65
Online exclusive
193.00
Save 7.35
Each
Add to cart
pNPP
may be used as a substrate for alkaline phosphatase (AP) in enzyme immunoassays.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Product Type
pNPP Solution
Quantity
100 ML
Shipping Condition
Wet Ice
Substrate
pNPP
Substrate Properties
Chemical Substrate
Substrate Type
AP (Alkaline Phosphatase) Substrate
Detection Method
Colorimetric
Unit Size
Each
Contents & Storage
Store at 2-8°C