Smart Diffuse Reflectance FT-IR Accessory
Actual product may vary
Thermo Scientific™

Smart Diffuse Reflectance FT-IR Accessory

Thermo Scientific™ Smart Diffuse Reflectance accessory™ maximizes the collection of diffusely scattered radiation, while minimizing the specular effect, resulting in the highest quality spectral results.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberFor Use With (Equipment)Type
0028-499Nicolet iS10 and Nicolet iS50Smart Diffuse Reflectance
0028-050Nicolet FT-IR InstrumentsSample Slide
0028-051Nicolet FT-IR InstrumentsReference Slide
0028-497Nicolet 380 and AvatarSmart Diffuse Reflectance
00284XXNicolet FT-IR InstrumentsSmart Diffuse Reflectance
Catalog number 0028-499
Price (MXN)
-
For Use With (Equipment):
Nicolet iS10 and Nicolet iS50
Type:
Smart Diffuse Reflectance
Thermo Scientific™ Smart Diffuse Reflectance accessory™ maximizes the collection of diffusely scattered radiation, while minimizing the specular effect, resulting in the highest quality spectral results. With the Smart Diffuse Reflectance accessory, a wide variety of samples can be analyzed with a minimum amount of sample preparation. With a slight amount of grinding and mixing with potassium bromide (KBr), samples of almost any type provide high quality spectra for qualitative or quantitative analysis.
Integral sample cups eliminate the need to worry about easily lost small parts, and are easy to load without the inconvenience of handling small sample cups. The accessory comes with two sample slides:
  • One has two integral sample cups within it - one position can be loaded with the sample and the other can be loaded with a reference material, such as KBr
  • Second holder has an integral gold mirror that can be used as a background reference and is ideal for use with the Si-Carb Sampling Kit

Specifications

Sample Cup Volume: 0.25g

Dual Sample Slide Holders: Si-Carb holder and powder sampler holder

KBr PowderPackets: 25 packets, 0.5g each

Silicon Carbide Disks: 320 and 400 grit Si-Carb adhesive-backed disks, 100 each

Includes:

Si-Carb Sampling Kit

  • Used to obtain spectra of hard, intractable samples , such as coatings, paints, and hard polymers
  • Includes adhesive-backed silicon carbide paper used to abrade the surface to be analyzed, and a handle and a sample plate to hold the paper
  • An infrared spectrum is obtained of the material clinging to the surface of the silicon carbide disk

KBr Powder Packets

  • KBr can be used to dilute samples prior to analysis
  • Contain a convenient, pre-measured quantity of dry KBr for dispersing a sample
Specifications
DescriptionThermo Scientific Smart Diffuse Reflectance accessory maximizes the collection of diffusely scattered radiation, while minimizing the specular effect, resulting in the highest quality spectral results.
For Use With (Application)Powders
IncludesDual Sample Slide Holders: Si-Carb Sampling Kit; KBr powder packs
OpticsGold-coated Compound Parabolic Concentrator
Volume (Metric)0.25 g
For Use With (Equipment)Nicolet iS10 and Nicolet iS50
TypeSmart Diffuse Reflectance
Unit SizeEach