Collector™ II Diffuse Reflectance Accessory
Imagem ilustrativa. O produto pode variar
Thermo Scientific™
Collector™ II Diffuse Reflectance Accessory
Thermo Scientific™ Collector II Diffuse Reflectance Accessory is designed for high sensitivity analysis powder samples and gems.
Have Questions?
Alterar visualizaçãobuttonViewtableView
Número do catálogoFor Use With (Equipment)Type
0030-150Spectra-Tech Collector IIDual Sample Cup Holder
0030-099Nicolet iS20 and Nicolet iS50Diffuse Reflectance Accessory
2001-106Collector IIMirror Adjustment Tool
3000-316Collector IISampling Cup, 0.25 g capacity
3000-319Collector IISampling Cup, 0.01 g capacity
3000-320Collector IIFunnel
7004-015Collector IIAlignment Mirror
840-224700Nicolet iS5, Summit SpecDiffuse Reflectance Accessory
Número do catálogo 0030-150
Preço (BRL)
-
For Use With (Equipment):
Spectra-Tech Collector II
Type:
Dual Sample Cup Holder
The Collector II is one of the most advanced and versatile accessories for Diffuse Reflectance Infrared analysis. It meets the needs of R and D laboratories that need a versatile sampling accessory.
Advantages
  • Rugged, high throughput optics provide high sensitivity for the analysis of powders
  • Patented Blocker device that removes specular reflected energy, allowing distortion-free diffuse reflection spectra
  • Integral purge allows removal of atmospheric interferences from carbon dioxide and water from spectral data
  • Easy, reproducible sample loading and protected optics
Environmental Chambers
  • Provide precise control of sample environment
  • Conditions that can be obtained include elevated temperatures or elevated or reduced pressure, non-ambient gaseous environments, and ambient environments
  • High Temperature/High Pressure Chamber or the High Temperature/Vacuum Chamber allow studies such as heterogeneous catalytic mechanisms, thermal degradation, zeolite analysis and hazardous environment
Includes:
Si-Carb Sample Prep Kit
  • Expands the analyses that can be performed beyond powders and small solid samples
  • Includes silicon-carbide disks used to abrade the sample
  • An infrared spectrum can be obtained of the material adhering to the surface of the disk
Recommended for:
  • Powders
  • Gems
  • Si-Carb scrapes
Especificações
Certifications/ComplianceISO 9001:2000
For Use With (Application)Reproducible Sample Loading
For Use With (Equipment)Spectra-Tech Collector II
TypeDual Sample Cup Holder
Unit SizeEach