Product Image

Digest-All 3 (Pepsin)

This product may be used to digest formalin-fixed, paraffin-embedded tissues (see Digest-All Kit above), and will improve the signal of certain antigens.
Have Questions?
Catalog NumberQuantity
00300950 mL
Catalog number 003009
Price (USD)
140.00
Each
Quantity:
50 mL
Price (USD)
140.00
Each

This product may be used to digest formalin-fixed, paraffin-embedded tissues (see Digest-All Kit above), and will improve the signal of certain antigens. It contains 50 mL of ready-to-use Pepsin. Digest-All™ 3 is stable for 12 months.

For Research Use Only. Not for use in diagnostic procedures.

Specifications
Product LineDigest-All
Quantity50 mL
Reagent TypeAntigen Retrieval Reagent
Shelf Life18 Months
Shipping ConditionWet Ice
TypeAntigen Retrieval Reagent
Unit SizeEach
Contents & Storage
Store in refrigerator (2–8°C).

Shelf life: 18 months from date of manufacture