Antibody Diluent
Actual product may vary
Thermo Scientific™

Antibody Diluent

Ready-to-use diluent with BSA (for stabilizing the antibody) and preservative.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
003218500 mL
003118250 mL
Catalog number 003218
Price (USD)
255.00
Each
Quantity:
500 mL
Price (USD)
255.00
Each

Ready-to-use diluent with BSA (for stabilizing the antibody) and preservative.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
FormLiquid
Product TypeAntibody Diluent
Shipping ConditionWet Ice
Quantity500 mL
Unit SizeEach
Contents & Storage
Store in refrigerator at 2°C to 8°C
  • Shelf life: 18 months from date of manufacture
  • Citations & References (1)

    Citations & References
    Abstract
    Carcinomas in situ of the breast with indeterminate features: role of E-cadherin staining in categorization.
    Authors:Jacobs TW, Pliss N, Kouria G, Schnitt SJ
    Journal:Am J Surg Pathol
    PubMed ID:11176072
    'Most breast carcinomas in situ (CIS) are easily categorized as ductal (DCIS) or lobular (LCIS). However, some CIS have indeterminate histologic features (CIS-IF). Prior studies have shown that E-cadherin protein expression is lost in lobular but not ductal carcinomas. Therefore, evaluation of examples of CIS-IF for E-cadherin expression by immunohistochemistry ... More