Variable Compensation Objectives
Actual product may vary
Thermo Scientific™
Variable Compensation Objectives
The Thermo Scientific™ Variable Compensation Objectives correct for spherical aberration caused when infrared windows are placed above the sample (up to 3mm in thickness), or when the Micro Compression Cell (Part No. 0045-434) is used.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescription
0045-40432X Infinity Corrected Objective
0042-40132X Reflachromat Objective
0042-50615X Reflachromat Objective
0045-40215X Infinity Corrected Objective
Catalog number 0045-404
Price (USD)
-
Description:
32X Infinity Corrected Objective
The Thermo Scientific™ Variable Compensation Objectives correct for spherical aberration caused when infrared windows are placed above the sample (up to 3mm in thickness), or when the Micro Compression Cell (Part No. 0045-434) is used.
Unique design for variable focal plane
Accommodates Slide-On ATR Objective accessory
Easily purged
15X Reflachromat Objective
0.58 N.A.
Working distance: up to 15mm (with the purge baffle removed)
Can also be used for ATR microscopy when used with the Slide-On ATR Attachment, purchased separately
32X Reflachromat Objective
0.65 N.A.
Working distance: 8mm Increased magnification makes it easier to define small sample areas
Recommended for:
Forensics
Pharmaceuticals
Polymers
Packaging
Specifications
Description32X Infinity Corrected Objective
For Use With (Application)Spectroscopy Elemental Isotope Analysis
For Use With (Equipment)Nicolet Continuum, Momentum
Unit SizeEach