15X Cassegrain Objectives (Fixed Compensation)
El producto real puede variar
Thermo Scientific™
15X Cassegrain Objectives (Fixed Compensation)
The Thermo Scientific™ 0.58 N.A., 15X Cassegrain objectives with fixed compensation come in three compensation configurations—zero, 1mm, or 2mm compensation objectives.
Have Questions?
Cambiar vistabuttonViewtableView
Número de catálogoCompensaciónPara utilizar con (equipo)
0047-4122 mmTodos los microscopios
0045-4000Nicolet Continuum, Momentum
0045-4352 mmNicolet Continuum, Momentum
0047-4100Todos los microscopios
0047-4111 mmMicroscopios de investigación IRus, Nic-Plan e IR-Plan
Número de catálogo 0047-412
Precio (CLP)
-
Compensación:
2 mm
Para utilizar con (equipo):
Todos los microscopios
The Thermo Scientific™ 0.58 N.A., 15X Cassegrain objectives with fixed compensation come in three compensation configurations—zero, 1mm, or 2mm compensation objectives.
The 0.58 N.A., 15X Fixed Cassegrain Objective has a working distance of 15mm. Objectives are available with three compensation configurations—zero, 1mm, and 2mm.The zero compensation objective is optimized for transmission/reflectance measurements when no window material is above the sample, the 1mm compensation objective is optimized for measurements when a 1mm thick window is above the sample, and the 2mm compensation objective is optimized for a 2mm window above the sample.
  • Prealigned objectives
  • Unique design for low cost and durability
  • Easily purged
Recommended for:
  • Forensics
  • Polymers
  • Pharmaceuticals
  • Packaging
Especificaciones
Compensación2 mm
Descripción15X Cassegrain objective; Fixed compensation; 2mm compensation
Para utilizar con (aplicación)Spectroscopy Elemental Isotope Analysis
Para utilizar con (equipo)Todos los microscopios
Unit SizeEach