Variable Compensation Condensers
Actual product may vary
Thermo Scientific™
Variable Compensation Condensers
The Thermo Scientific™ Variable Compensation Condensers correct for spherical aberration caused by infrared windows beneath the sample (up to 3mm in thickness).
Have Questions?
보기 방식 변경buttonViewtableView
카탈로그 번호설명
0047-41815X Condenser with Variable Compensation
0042-50710X Condenser
0045-40315X Infinity Corrected Condenser
0045-40532X Infinity Corrected Condenser
0047-41932X Condenser with Variable Compensation
카탈로그 번호 0047-418
제품 가격(KRW)
-
설명:
15X Condenser with Variable Compensation
The Thermo Scientific™ Variable Compensation Condensers correct for spherical aberration caused by infrared windows beneath the sample (up to 3mm in thickness).
Variable compensation
Easily purged
10X Reflachromat Condenser
0.71 N.A.
Standard condenser for all IRμs, Nic-Plan and IR-Plan research microscopes (not available for any other microscopes)
15X Reflachromat Condenser
0.58 N.A.
Corrects for spherical aberration caused by infrared windows beneath the sample (up to 3mm in thickness)
15X Reflachromat Condenser
0.65 N.A.
Increased magnification allows it to be matched to the objective
Recommended for:
Textiles
Polymers
Pharmaceuticals
Packaging
Forensic
사양
보상Variable
설명15X Condenser with Variable Compensation
용도(애플리케이션)Spectroscopy Elemental Isotope Analysis
용도 (장비)IRus, Nic-Plan, IR-Plan Research Microscopes
유형Condenser
Unit SizeEach