Citrate Buffer (pH 6.0), Concentrate - FAQs
View additional product information for Citrate Buffer (pH 6.0), Concentrate - FAQs (005000)
1 product FAQs found
What is the concentration/molarity of the Citrate Buffer (pH 6.0), Concentrate (Cat. No. 005000)?
The Citrate Buffer (pH 6.0), Concentrate (Cat. No. 005000) is a 20X solution and contains two compounds with different molecular weights, so there is no single molarity for the formulation. The citric acid is at about 20 mM and sodium citrate is at about 180 mM.
Find additional tips, troubleshooting help, and resources within our Cell Imaging Support Center.