B-27™ Supplement (50X), custom
Gibco™

B-27™ Supplement (50X), custom

The custom B-27 Supplement (50X) is a serum-free additive for the growth and expansion of retinal ganglion cells.
Have Questions?
Catalog NumberQuantity
0080085SA10 mL
Catalog number 0080085SA
Price (USD)
219.00
Each
Quantity:
10 mL
Price (USD)
219.00
Each

The custom B-27 Supplement (50X) is a serum-free additive for the growth and expansion of retinal ganglion cells. It is supplied as a 50X liquid concentrate and it is used with a basal medium, such as Neurobasal, and other additives, such as GlutaMAX-1.

For Research Use Only. Not for use in diagnostic procedures.

Specifications
Cell TypeNeural Cells
Concentration50 X
Culture TypeMammalian Cell Culture
Endotoxin Level≤ 25 EU/ml
Product LineGibco
Purity or Quality GradeCell Culture Grade
Quantity10 mL
Serum LevelSerum-Free
Shipping ConditionDry Ice
SourceSerum-Free
Tested ForSterility Testing (Bacteria & Fungi), Endotoxin (EU/ml), Growth Performance Assay
ClassificationSerum-free
FormLiquid
Product TypeCell Culture Supplement
SterilitySterile
With AdditivesAntioxidant Cocktail, Insulin, Vitamin A
pH6 to 8
Unit SizeEach
Contents & Storage
10 mL - 50X frozen concentrate; store at -5°C to -20°C in the dark.

Frequently asked questions (FAQs)

Do you offer B-27 supplements with and without insulin?

Yes, we do offer B-27 Supplement, minus insulin (Cat. Nos. A1895601 and A1895602). All our other B-27 supplements do contain insulin.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.