Product Image
Invitrogen™
SPoT-Light Tissue Pretreatment Kit
For Enzyme and Heat Pretreatment of FFPE Tissue prior to CISH.
Have Questions?
Catalog NumberQuantity
008401
also known as 00-8401
1 Kit
Catalog number 008401
also known as 00-8401
Price (USD)
265.00
Each
Quantity:
1 Kit
Price (USD)
265.00
Each
For Enzyme and Heat Pretreatment of FFPE Tissue prior to CISH.
For Laboratory Use
Specifications
Product LineCISH, SPOT-Light
Quantity1 Kit
Reagent TypeAntigen Retrieval Reagent
Shelf Life18 Months
Shipping ConditionWet Ice
TypeTissue Pretreatment Kit
Unit SizeEach
Contents & Storage
Store in refrigerator (2–8°C).
Shelf life: 18 months from date of manufacture
Citations & References (1)
Citations & References
Abstract
EGFR status in oral squamous cell carcinoma: comparing immunohistochemistry, FISH and CISH detection in a case series study.
Authors:Bernardes VF, Gleber-Netto FO, Sousa SF, Rocha RM, Aguiar MC,
Journal:BMJ Open
PubMed ID:23358562
To compare the immunohistochemistry (IHC) expression of epidermal growth factor receptor (EGFR) in oral squamous cell carcinomas (OSCC) with the gene amplification evaluated by fluorescence in situ hybridization (FISH) and chromogenic in situ hybridization (CISH) and their association with clinicopathological parameters. Additionally, we tested the sensibility and specificity of CISH ... More