Molybdenum foil, 0.1mm (0.004in) thick, 99.95% (metals basis)
Molybdenum foil, 0.1mm (0.004in) thick, 99.95% (metals basis)
Molybdenum foil, 0.1mm (0.004in) thick, 99.95% (metals basis)
Thermo Scientific Chemicals

Molybdenum foil, 0.1mm (0.004in) thick, 99.95% (metals basis)

Form: approx. 10.2g/100x100mm | CAS: 7439-98-7 | Mo | 95.95 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 x 150 mm
150 x 300 mm
300 x 600 mm
Catalog number 010043.GJ
also known as 010043-GJ
Price (USD)
63.65
Special offer
Online exclusive
Ends: 30-Jun-2026
75.20
Save 11.55 (15%)
Each
Quantity:
100 x 150 mm
Request bulk or custom format
Price (USD)
63.65
Special offer
Online exclusive
Ends: 30-Jun-2026
75.20
Save 11.55 (15%)
Each
Chemical Identifiers
CAS7439-98-7
IUPAC Namemolybdenum
Molecular FormulaMo
InChI KeyZOKXTWBITQBERF-UHFFFAOYSA-N
SMILES[Mo]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Grey
Assay from Supplier's CofA≥99.95% (metals basis)
Assay from Supplier's CofACr: ≤ 0.005 %
Assay from Supplier's CofACu: ≤ 0.002 %
Assay from Supplier's CofAMg: ≤ 0.001 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only