Molybdenum foil, 1.0mm (0.04in) thick, 99.95% (metals basis)
Molybdenum foil, 1.0mm (0.04in) thick, 99.95% (metals basis)
Molybdenum foil, 1.0mm (0.04in) thick, 99.95% (metals basis)
Thermo Scientific Chemicals

Molybdenum foil, 1.0mm (0.04in) thick, 99.95% (metals basis)

CAS: 7439-98-7 | Mo | 95.95 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 x 50 mm
100 x 100 mm
200 x 200 mm
Catalog number 010046.FI
also known as 010046-FI
Price (USD)
67.65
Special offer
Online exclusive
Ends: 30-Jun-2026
79.40
Save 11.75 (15%)
Each
Quantity:
50 x 50 mm
Request bulk or custom format
Price (USD)
67.65
Special offer
Online exclusive
Ends: 30-Jun-2026
79.40
Save 11.75 (15%)
Each
Chemical Identifiers
CAS7439-98-7
IUPAC Namemolybdenum
Molecular FormulaMo
InChI KeyZOKXTWBITQBERF-UHFFFAOYSA-N
SMILES[Mo]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities≤0.05%
FormFoil, 1.0 mm (0.04 in.) thick
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only