Platinum Iridium wire, 0.5mm (0.02in) dia, hard
Thermo Scientific Chemicals

Platinum Iridium wire, 0.5mm (0.02in) dia, hard

Form: Approx. 0.44g/10cm, Temper: as drawn | CAS: 7440-6-6 | Pt:Ir; 90:10 wt%
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 cm
50 cm
250 cm
Catalog number 010056.CB
also known as 010056-CB
Price (USD)
-
Quantity:
250 cm
Request bulk or custom format
Specifications
AppearanceSilvery white wire
CAS6-6-7440
Chemical Name or MaterialPlatinum
ColorWhite
MDL NumberMFCD02091683
View more
Fabrication of metal microelectrodes for electrical stimulation of nervous tissue and electrophysiological recordings. Inoculating needles and leadwires for implants. Electrode material for gel boxes/electrophoresis chambers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only