Platinum 30 wt% Rhodium wire, 0.25mm (0.01 in.) dia., ISA Type B Standard Grade Thermocouple, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Platinum 30 wt% Rhodium wire, 0.25mm (0.01 in.) dia., ISA Type B Standard Grade Thermocouple, Thermo Scientific Chemicals

For ISA Type B Standard Grade Thermocouple
Have Questions?
Catalog NumberQuantity
010063.BY
also known as 010063-BY
100 cm
Catalog number 010063.BY
also known as 010063-BY
Price (USD)
-
Quantity:
100 cm
Request bulk or custom format
Specifications
Avg. Mol. Wt. or Mol. Wt. RangePt:Rh; 70:30 wt%
Chemical Name or MaterialPlatinum 30 wt% Rhodium wire
MDL NumberMFCD00198186
Name Note0.25mm (0.01 in.) dia.
OdorOdorless
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only