Cadmium rod, 12.7mm (0.5in) dia x 23cm (9.0in), Puratronic™, 99.999+% (metals basis)
Cadmium rod, 12.7mm (0.5in) dia x 23cm (9.0in), Puratronic™, 99.999+% (metals basis)
Cadmium rod, 12.7mm (0.5in) dia x 23cm (9.0in), Puratronic™, 99.999+% (metals basis)
Thermo Scientific Chemicals

Cadmium rod, 12.7mm (0.5in) dia x 23cm (9.0in), Puratronic™, 99.999+% (metals basis)

CAS: 7440-43-9 | Cd | 112.41 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 Pc.
5 Pc.
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 010120.KG
also known as 010120-KG
Price (USD)
-
Quantity:
1 Pc.
Request bulk or custom format
Chemical Identifiers
CAS7440-43-9
IUPAC Namecadmium
Molecular FormulaCd
InChI KeyBDOSMKKIYDKNTQ-UHFFFAOYSA-N
SMILES[Cd]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White silvery
FormRod, 12.7mm (0.5in) diameter.
Assay from Supplier's CofA>99.999% (metals basis)
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov
RUO – Research Use Only