Germanium ingot, zone refined, 99.9999% (metals basis), electrical resistivity: 40 ohm-cm
Germanium ingot, zone refined, 99.9999% (metals basis), electrical resistivity: 40 ohm-cm
Germanium ingot, zone refined, 99.9999% (metals basis), electrical resistivity: 40 ohm-cm
Germanium ingot, zone refined, 99.9999% (metals basis), electrical resistivity: 40 ohm-cm
Thermo Scientific Chemicals

Germanium ingot, zone refined, 99.9999% (metals basis), electrical resistivity: 40 ohm-cm

Form: Electrical Resistivity: 40 ohm-cm | CAS: 7440-56-4 | Ge | 72.63 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
24 g
120 g
Catalog number 010192.79
also known as 010192-79
Price (USD)
1,521.65
Online Exclusive
1,690.00
Save 168.35 (10%)
Each
Add to cart
Quantity:
120 g
Request bulk or custom format
Price (USD)
1,521.65
Online Exclusive
1,690.00
Save 168.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS7440-56-4
IUPAC Namegermanium
Molecular FormulaGe
InChI KeyGNPVGFCGXDBREM-UHFFFAOYSA-N
SMILES[Ge]
View more
SpecificationsSpecification SheetSpecification Sheet
FormIngot
Appearance (Color)Gray to white
Total Metal Impurities0.0001% max.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only