Holmium foil, 0.3mm (0.01in) thick, 99.9% (REO)
Holmium foil, 0.3mm (0.01in) thick, 99.9% (REO)
Holmium foil, 0.3mm (0.01in) thick, 99.9% (REO)
Thermo Scientific Chemicals

Holmium foil, 0.3mm (0.01in) thick, 99.9% (REO)

CAS: 7440-60-0 | Ho | 164.93 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 x 25 mm
50 x 50 mm
Catalog number 010210.FI
also known as 010210-FI
Price (USD)
880.65
Online Exclusive
978.00
Save 97.35 (10%)
Each
Quantity:
50 x 50 mm
Request bulk or custom format
Price (USD)
880.65
Online Exclusive
978.00
Save 97.35 (10%)
Each
Chemical Identifiers
CAS7440-60-0
IUPAC Nameholmium
Molecular FormulaHo
InChI KeyKJZYNXUDTRRSPN-UHFFFAOYSA-N
SMILES[Ho]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Rare Earth Oxide Impurities0.1% max.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
  • Packaged under argon
RUO – Research Use Only