Thermo Scientific Chemicals

Lead powder, -200 mesh, 99.9% (metals basis), Thermo Scientific Chemicals

產品號碼: 010223.A1
1 kg, Each
Thermo Scientific Chemicals

Lead powder, -200 mesh, 99.9% (metals basis), Thermo Scientific Chemicals

產品號碼: 010223.A1
1 kg, Each
Quantity
產品號碼: 010223.A1
亦稱為 010223-A1
價格 (TWD)

化學識別

CAS
7439-92-1
IUPAC Name
lead
Molecular Formula
Pb
InChI Key
WABPQHHGFIMREM-UHFFFAOYSA-N
SMILES
[Pb]
Total Metal Impurities
0.1% max.

說明

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

圖表

文件與下載

證書

    常見問答集 (常見問題)

    引用資料與參考文獻

    Search citations by name, author, journal title or abstract text