Magnesium powder, -325 mesh, 99.8%, Thermo Scientific Chemicals
Magnesium powder, -325 mesh, 99.8%, Thermo Scientific Chemicals
Magnesium powder, -325 mesh, 99.8%, Thermo Scientific Chemicals
Magnesium powder, -325 mesh, 99.8%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Magnesium powder, -325 mesh, 99.8%, Thermo Scientific Chemicals

CAS: 7439-95-4 | Molecular Formula: Mg | Molecular Weight: 24.30 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
100 g
500 g
4 x 500 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 010233.22
Price (USD)
-
Quantity:
100 g
Request bulk or custom format
Chemical Identifiers
CAS7439-95-4
IUPAC Namemagnesium
Molecular FormulaMg
InChI KeyJLVVSXFLKOJNIY-UHFFFAOYSA-N
SMILES[Mg]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Metallic white to grey
FormPowder
Assay≥99.8%
Particle Size-325 Mesh

Poly(vinylidene fluoride) is useful in coatings, film, filter cloth, instrument linings, filtration membranes, pump parts, linings and valves. It is also employed as a material for transducers in devices such as headphones, microphones and sonic detectors. It is also essential for piezoelectric and electrostrictive applications. In addition, it is useful for monofilament fishing lines replacing nylon monofilament. It acts as a standard binder used in the production of composite electrodes for lithium ion batteries. It serves as a high grade insulator for wires, nanomaterials and pressure sensors. It is utilized to make filters which are used to prepare samples for high performance liquid chromatography and other advanced analytical techniques.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

RUO – Research Use Only