Nickel wire, 0.38mm (0.015 in.) dia., 99.8% (metals basis), Thermo Scientific Chemicals
Nickel wire, 0.38mm (0.015 in.) dia., 99.8% (metals basis), Thermo Scientific Chemicals
Nickel wire, 0.38mm (0.015 in.) dia., 99.8% (metals basis), Thermo Scientific Chemicals
Nickel wire, 0.38mm (0.015 in.) dia., 99.8% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Nickel wire, 0.38mm (0.015 in.) dia., 99.8% (metals basis), Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantity
010253.H4
also known as 010253-H4
100 m
Catalog number 010253.H4
also known as 010253-H4
Price (USD)
130.65
Special offer
Online exclusive
Ends: 30-Jun-2026
154.00
Save 23.35 (15%)
Each
Quantity:
100 m
Request bulk or custom format
Price (USD)
130.65
Special offer
Online exclusive
Ends: 30-Jun-2026
154.00
Save 23.35 (15%)
Each
Chemical Identifiers
CAS7440-02-0
IUPAC Namenickel
Molecular FormulaNi
InChI KeyPXHVJJICTQNCMI-UHFFFAOYSA-N
SMILES[Ni]
View more
SpecificationsSpecification SheetSpecification Sheet
FormWire, 0.38mm (0.015in) diameter.
Appearance (Color)Silvery
Assay from Supplier's CofA≥99.8% (metals basis)
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only