Niobium foil, 1.0mm (0.04in) thick, annealed, 99.8% (metals basis)
Niobium foil, 1.0mm (0.04in) thick, annealed, 99.8% (metals basis)
Niobium foil, 1.0mm (0.04in) thick, annealed, 99.8% (metals basis)
Thermo Scientific Chemicals

Niobium foil, 1.0mm (0.04in) thick, annealed, 99.8% (metals basis)

Form: Approx.42.9g/50 x 100mm | CAS: 7440-03-1 | Nb | 92.91 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 x 100 mm
100 x 200 mm
200 x 500 mm
Catalog number 010257.GK
also known as 010257-GK
Price (USD)
289.65
Special offer
Online exclusive
Ends: 30-Jun-2026
340.00
Save 50.35 (15%)
Each
Quantity:
100 x 200 mm
Request bulk or custom format
Price (USD)
289.65
Special offer
Online exclusive
Ends: 30-Jun-2026
340.00
Save 50.35 (15%)
Each
Chemical Identifiers
CAS7440-03-1
IUPAC Nameniobium
Molecular FormulaNb
InChI KeyGUCVJGMIXFAOAE-UHFFFAOYSA-N
SMILES[Nb]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.2% max.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only