Niobium foil, 0.127mm (0.005in) thick, annealed, 99.8% (metals basis)
Niobium foil, 0.127mm (0.005in) thick, annealed, 99.8% (metals basis)
Niobium foil, 0.127mm (0.005in) thick, annealed, 99.8% (metals basis)
Thermo Scientific Chemicals

Niobium foil, 0.127mm (0.005in) thick, annealed, 99.8% (metals basis)

CAS: 7440-03-1 | Nb | 92.91 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 x 10 cm
10 x 20 cm
20 x 50 cm
Catalog number 010261.CS
also known as 010261-CS
Price (USD)
99.65
Online Exclusive
110.00
Save 10.35 (9%)
Each
Quantity:
10 x 20 cm
Request bulk or custom format
Price (USD)
99.65
Online Exclusive
110.00
Save 10.35 (9%)
Each
Chemical Identifiers
CAS7440-03-1
IUPAC Nameniobium
Molecular FormulaNb
InChI KeyGUCVJGMIXFAOAE-UHFFFAOYSA-N
SMILES[Nb]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities≤0.2%
FormFoil, 0.127mm (0.005in) thick, annealed
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only